missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEF2 Recombinant Rabbit Monoclonal Antibody (23GB1550)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | EEF2 |
|---|---|
| Clone | 23GB1550 |
| Concentration | 0.33 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
EEF2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.Specifications
| EEF2 | |
| 0.33 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05197, P13639, P58252 | |
| 13629, 1938, 29565 | |
| A synthesized peptide derived from human EEF2 (800-858AA). | |
| Antibody |
| 23GB1550 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| EEF2 | |
| EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2 | |
| EEF2 | |
| Primary | |
| Affinity chromatography |