Edwards & Cromwell Universal Leak Control, AE Series Kits DFS Item
$2,380.54 - $3,284.74
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
19-811-089 DFS Item
View Documents Promotion Details
Edwards & Cromwell
AENS
With non-sparking tools
Each for $3,284.74
19-823-752 DFS Item
View Documents Promotion Details
Edwards & Cromwell
AE
With steel tools
Each for $2,380.54
Description
Description
Kit contains commonly used components for emergency and temporary leak stoppage of liquid or bulk hazardous materials due to punctures, gashes, cracks, or surface rotting.
  • Designed for small to medium holes in any type of low-pressure vessel
  • Covers pin, convex, concave, and flat surface holes as well as cracks of all shapes
  • Employs quick application, twin “T” patch and ladder patch design for average forklift-type punctures
These kits designed for use only by experienced and trained personnel only for temporary control until the container can be disposed of properly. Proper protective clothing must be worn.