Please login to your online account to display your discounted pricing
Edvotek Quick Plants™ Seeds
From seed to flower in 6-7 weeks.
$29.70 - $35.10
Specifications
For Use With (Application) Rapid growth make these plants ideal for classroom use.
View More Specs
Products
Catalog Number Mfr. No. Scientific Name Product Type Quantity Price Quantity    
S11951
fisher science education
1251
Arabidopis thaliana Wild Type Quick Plant Seed 150 Seeds Each for $29.70
S11952
fisher science education
1252
Arabidopis thaliana Wild Type Quick Plant Seed 300 Seeds Each for $35.10
S11957
fisher science education
1257
Arabidopis thaliana Variegated Quick Plant Seed 150 Seeds Each for $30.05
S11958
fisher science education
S11958
Arabidopis thaliana Variegated Quick Plant Seed 300 Seeds Pack of 300 Each for $35.10
Description & Specifications
Specifications
For Use With (Application) Rapid growth make these plants ideal for classroom use.
  • Grow rapidly and flourish under fluorescent light
  • Rapid growth make these plants ideal for classroom use