Please login to your online account to display your discounted pricing

Edvotek™ M12 and M12 Dual Electrophoresis Apparatus


Host two lab groups on one apparatus.

Manufacturer: fisher scientific  502*

 View more versions of this product

Catalog No. S11502

  • / Each
 

Description & Specifications

Specifications

Warranty Lifetime
Volume (Metric) Buffer 400mL
Length (Metric) Gel 14cm
Width (English) Gel 2.7 in.
Width (Metric) Gel 7cm
Volume (English) Buffer 13.5 oz.
Length (English) Gel 5.5 in.
Analysis Time 30 to 40 min.
Includes One 7 x 14cm trays, two 6-teeth combs, one 8/10-teeth comb, two Rubber end caps
Model M12 single
Sample Size Up to 12
Product Type M12 single Electrophoresis Apparatus