Please login to your online account to display your discounted pricing

Edvotek™ M12 and M12 Dual Electrophoresis Apparatus

Host two lab groups on one apparatus.

$369.10 - $379.35

Specifications

Analysis Time 30 to 40 min.
Volume (Metric) Buffer 400mL
Width (Metric) Gel 7cm
Warranty Lifetime
View More Specs
Includes:  S90242ND
  • Two 7 × 7cm trays
  • Two 6-tooth combs
  • Two 8-/10-tooth combs
  • Four rubber end caps
S11502
  • One 7 × 14cm tray
  • Two 6-tooth combs
  • One 8-/10-tooth comb
  • Two rubber end caps
Products
Catalog Number Mfr. No. Includes Model Sample Size Price Quantity    

S11502

 
fisher scientific
502*
One 7 x 14cm trays, two 6-teeth combs, one 8/10-teeth comb, two Rubber end caps M12 single Up to 12 Each for $369.10

S90242ND

 
fisher scientific
504*
One 7 x 14cm trays, two 6-teeth combs, one 8/10-teeth comb, four Rubber end caps M12 dual Up to 20 Each for $379.35
Description & Specifications

Specifications

Analysis Time 30 to 40 min.
Volume (Metric) Buffer 400mL
Width (Metric) Gel 7cm
Warranty Lifetime

Choose either two 7 × 7cm gel trays (dual model) or one 7 x 14cm gel tray (single model).

  • New, sleek design
  • Contoured lid enhances visualization
  • Vented base
  • Color coded push tabs
  • Pour spout for disposing of used buffer
  • Replaceable electrode modules
  • Results in 30-40 minutes
  • Patent pending