Edvotek™ DNA Fingerprinting: Identification of DNA by Restriction Fragmentation Patterns
Reveals techniques used in forensic and medical procedures.
Supplier:  Edvotek™ EMD109
Catalog No. S11109
Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, FlashBlue™ DNA Stain, InstaStain™ Blue Cards, & Disposable Pipets.
Requires : Horizontal gel electrophoresis apparatus; DC power supply; Staining tray; Visualization system (white light); Micropipets with tips to obtain samples for five gels; 250mL flasks or beakers; Distilled or deionized water; Hotplate, Bunsen burner or microwave; Balance
Description
Description
Basic concepts of DNA fingerprinting are featured in this lab by comparing crime scene DNA with suspect DNAs. Fingerprint patterns are separated by agarose gel electrophoresis and the students determine who may have done-it!
Specifications
Specifications
8 Gels