Please login to your online account to display your discounted pricing

EDTA Titrant, 0.0250 Molar, Ricca Chemical

$177.88 - $177.88

Chemical Identifiers

Chemical Name or Material EDTA Titrant
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2703-1

  SDS
ricca chemical company
RI27031
Poly natural 4L Each for $177.88

2703-16

  SDS
ricca chemical company
RI270316
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2703-32

 
ricca chemical company
RI270332
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2703-5

 
ricca chemical company
RI27035
Cubitainer™ 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R270300010F

 
ricca chemical company
RIR270300010F
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 0.0250Molar
Chemical Name or Material EDTA Titrant
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural