missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDTA Titrant, 0.0200 Molar, Ricca Chemical

$30.06 - $150.97

Chemical Identifiers

CAS 7732-18-5,6381-92-6
CAS Min % 97.0,0.72
CAS Max % 100.0,0.75
Physical Form Liquid
Chemical Name or Material EDTA Titrant
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2702-32 Ricca Chemical Company
RI270232
Poly natural 1L Each for $51.97
2702-1 Ricca Chemical Company
RI27021
Poly natural 4L Each for $150.97
2702-5 Ricca Chemical Company
RI27025
Cubitainer™ 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R270200010F Ricca Chemical Company
RIR270200010F
Cubitainer 10L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R2702000500 Ricca Chemical Company
RIR2702000500A
Poly natural 500mL Each for $30.06
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6381-92-6
100.0,0.75
EDTA Titrant
97.0,0.72
Liquid

Specifications

Liquid
1L
97.0,0.72
Poly natural
EDTA Titrant
Clear colorless
0.0200Molar
100.0,0.75
6381-92-6,7732-18-5