missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EDTA Titrant, 0.00100 M (M/1000), 1 mL = 0.1 mg CaCO3 (0.04 mg Ca), Ricca Chemical
Supplier: Ricca Chemical Company R26920004A
Specifications
EDTA Titrant | |
6381-92-6,7732-18-5 | |
100.0,0.04 | |
MFCD00150037,MFCD00003541 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
1 mL = 0.1 mg CaCO | |
Natural Poly Bottle | |
Liquid |
Colorless | |
98.0,0.04 | |
C10H18N2Na2O10 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
44120005 | |
4 L | |
0.00100Molar |
Chemical Identifiers
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |