missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDTA Titrant, 0.00100 Molar, 1mL = 0.1mg CaCO, Ricca Chemical

$44.90 - $128.15

Chemical Identifiers

CAS 7732-18-5,6381-92-6
CAS Min % 98.0,0.04
CAS Max % 100.0,0.04
Physical Form Liquid
Chemical Name or Material EDTA Titrant
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2692-32 Ricca Chemical Company
RI269232
Poly natural 1L Each for $44.90
R269200010F Ricca Chemical Company
RIR269200010F
Cubitainer 10L Each for $128.15
2692-16 Ricca Chemical Company
RI269216
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R269200020F Ricca Chemical Company
RIR269200020F
Cubitainer 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R26920004A Ricca Chemical Company
RIR26920004A
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6381-92-6
100.0,0.04
EDTA Titrant
98.0,0.04
Liquid

Specifications

7732-18-5,6381-92-6
100.0,0.04
1 mL = 0.1 mg CaCO
0.00100Molar
1L
98.0,0.04
Liquid
Poly natural
Clear colorless
EDTA Titrant