missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EDTA Stabilizer, 50% (w/v) EDTA and 10% (w/v) NaOH, Ricca Chemical
For Ammonia Nitrogen Determination
Supplier: Ricca Chemical Company 268016
Specifications
EDTA Stabilizer | |
30.64,61.0,6.13 | |
C10H18N2Na2O10 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
Natural Poly Bottle | |
50% (w/v) EDTA + 10% (w/v) NaOH |
6381-92-6,7732-18-5,1310-73-2 | |
31.89,64.0,6.38 | |
MFCD00150037,MFCD00003541 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
131664146 | |
500 mL |