missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDTA Reagent, For Nitrogen (Ammonia), Nitrogen (Organic), 5% (w/v), LabChem™

Supplier:  LabChem LC139302

 View more versions of this product

Catalog No. LC139302


Description

Description

Specifications

Specifications

EDTA Reagent
For Nitrogen (Ammonia), Nitrogen (Organic)
5,94.95,0.05
(HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
131664146
5% (w/v)
Poly Bottle
1 L
Colorless
6381-92-6,7732-18-5,1310-73-2
C10H18N2Na2O10
MFCD00150037,MFCD00003541
Soluble in water
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.23
Passes Test
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Liquid

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
131664146
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions