missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDTA Disodium Magnesium Salt, Tetrahydrate, J.T.Baker™

$6,144.52 - $9,887.31

Chemical Identifiers

CAS 29932-54-5
Formula Weight 430.56
Chemical Name or Material EDTA Disodium Magnesium Salt, Tetrahydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02002783 Avantor Performance
H71006
Poly Pail 5kg Each for $9,887.31
02002782 Avantor Performance
H71005
Poly Bottle 100g Case of 12 Each for $6,144.52
Specifications Description & Specifications

Chemical Identifiers

29932-54-5
430.56
EDTA Disodium Magnesium Salt, Tetrahydrate

Specifications

29932-54-5
Poly Pail
EDTA Disodium Magnesium Salt, Tetrahydrate
430.56
5kg