Please login to your online account to display your discounted pricing

EDTA Disodium, Dihydrate, ACS Grade, 99.0 to 101.0%, LabChem™

$106.29 - $188.23

Chemical Identifiers

Chemical Name or Material EDTA Disodium
CAS 6381-92-6
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Formula Weight 372.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC137501

 
lab chem
LC137501
Poly Bottle 500g Each for $188.23

LC137507

 
lab chem
LC137507
Poly Bottle 100g Each for $106.29
Description & Specifications

Specifications

Formula Weight 372.23
Solubility Information Soluble in water
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
EINECS Number 205-358-3
Melting Point 252°C
Chemical Name or Material EDTA Disodium
Assay 99.0 to 101.0%
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
CAS 6381-92-6
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Purity Grade ACS Grade
CAS Max % 100
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Keep container tightly closed.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Physical Form Crystals
Name Note Dihydrate
Quantity 500g
Identification Passes Test
Health Hazard 1 Warning
Color White
Recommended Storage Room Temperature
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Keep container tightly closed.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature