missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EDTA (Disodium), 2.5% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,6381-92-6
CAS Min % 96.0,2.39
CAS Max % 100.0,2.49
Physical Form Liquid
Chemical Name or Material EDTA, Disodium
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
2665-16 Ricca Chemical Company
RI266516
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2665-32 Ricca Chemical Company
RI266532
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6381-92-6
100.0,2.49
EDTA, Disodium
96.0,2.39
Liquid

Specifications

7732-18-5,6381-92-6
100.0,2.49
Poly natural
Aqueous Solution
500mL
96.0,2.39
Liquid
2.5% (w/v)
Clear colorless
EDTA, Disodium