Please login to your online account to display your discounted pricing

EDTA Decontaminating Solution, For Radium, LabChem™

$80.09 - $80.09

Chemical Identifiers

Chemical Name or Material EDTA Decontaminating Solution
CAS 7732-18-5
CAS Max % 98
CAS 497-19-8
CAS Max % 1
CAS 6381-92-6
CAS Max % 1
Name Note For Radium
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Formula Weight 372.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC138002

 
lab chem
LC138002
1L Each for $80.09
Description & Specifications

Specifications

Formula Weight 372.23
Quantity 1L
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Color Clear, colorless
CAS Max % 1
Solubility Soluble in water
Identification Passes Test
Packaging Poly Bottle
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Physical Form Liquid
CAS Max % 98
Storage Information Room Temperature
CAS 497-19-8
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
CAS 7732-18-5
CAS 6381-92-6
CAS Max % 1
Chemical Name or Material EDTA Decontaminating Solution
Density 1g/mL
Name Note For Radium
Specific Gravity 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.