Please login to your online account to display your discounted pricing

EDTA, Certified, 0.0050M ±0.0001M (0.01N), LabChem™

$56.00 - $56.00

Chemical Identifiers

Chemical Name or Material EDTA
CAS 7732-18-5
CAS Max % 99.81
CAS 6381-92-6
CAS Max % 0.19
Purity Grade Certified Grade
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Formula Weight 372.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC138202

 
lab chem
LC138202
Poly Bottle 1L Each for $56.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 372.23
Chemical Name or Material EDTA
Density 1g/mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1g/mL
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
CAS 7732-18-5
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Concentration or Composition (by Analyte or Components) 0.0050M ±0.0001M (0.01N)
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Color Clear, colorless
Quantity 1L
CAS Max % 99.81
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 6381-92-6
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.19
Recommended Storage Room Temperature
Identification Passes Test
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature