Please login to your online account to display your discounted pricing

Edetate Disodium U.S.P., Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  139506

 View more versions of this product

Catalog No. MK1395212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 372.24
Quantity 2.5kg
Chemical Name or Material Edetate Disodium
Purity Grade USP
Molecular Formula C10H14N2Na2O8·2H2O
CAS 6381-92-6
Packaging Poly

Specifications

Chemical Name or Material Edetate Disodium
CAS 6381-92-6
Purity Grade USP
Molecular Formula C10H14N2Na2O8·2H2O
Formula Weight 372.24