Please login to your online account to display your discounted pricing

Edetate Disodium U.S.P. GenAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Edetate Disodium
CAS 6381-92-6
Purity Grade USP
Molecular Formula C10H14N2Na2O8·2H2O
Formula Weight 372.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7727500

 
avantor performance
772704
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 372.24
Chemical Name or Material Edetate Disodium
Purity Grade USP
Molecular Formula C10H14N2Na2O8·2H2O
CAS 6381-92-6
Packaging POLYSTORMOR
Quantity 500g