Please login to your online account to display your discounted pricing

Economy Ultraviolet Goggle Sanitizers


Sanitize and store 30 to 40 goggles or safety glasses in the same cabinet.

Manufacturer: fisher scientific  K50EMD

 View more versions of this product

Catalog No. S47604A

  • / Each
 

Description & Specifications

Specifications

Depth (English) 9.25 in.
For Use With (Equipment) 30 pairs of goggles
Includes timer
Type Goggle Sanitizer
Certifications/Compliance UL Approved
Device Holds 30 pairs
For Use With (Application) Sanitize, store goggles or safety glasses in the same cabinet
Height (English) 26 in.
Width (English) 24.5 in.