Economy Micropipetters
Achieve optimal pipetting. Poulten Graf™ Economy Micropipettes are used for measuring and transferring quantities of liquids from one container to another.
Manufacturer:  Fisher Scientific E100010
 View more versions of this product
Catalog No. S317184
Description
Description
Price breakthrough in micropipetters!
Unlike most micropipetters costing hundreds of dollars, these economy micropipetters relieve strained budgets.
•    Working accuracy of ±0.75 or better
•    Normal repeatability between 0.07% and 0.34%
•    Lightweight for easy pipetting
•    Tip ejector designed for large or small, right or left hands
•    All four models are continuously variable within their range
For pipetting into or out of test tubes, choose the blue model (extended shaft).
Specifications
Specifications
±0.75%
Turquoise
1000 to 10000μL
Documents