missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Troemner™ Economical Stainless Steel Weight Set, 2000g-10mg, Class 7, No Certificate,

For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.

Manufacturer:  Troemner™ 33810S

Catalog No. 19-216-360



Description

Description

  • Set of 22 Class 7 stainless steel weights
  • Supplied in a rugged, plastic case with a locking latch and compartment for each weight in the case
  • Manufactured in the United States
  • Does not include a NVLAP Accreditation Certificate
Specifications

Specifications

No Certificate
Weights above 5 g are 303 Stainless Steel. Weights 5 g and below are 3003-H14 Aluminum.
2000 g to 10 mg
Cylindrical with handling knob (weights 2000 g - 10 g), Flat with one end turned up (weights 5 g - 10 mg). Two pieces (weights 200 g or more), One piece (under 200 g).
Weights: 2000g, 1000g, 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg
Class 7
Rugged, plastic case with a locking latch and compartment for each weight in the case
Weights above 5g are 7.85g/cm3 at 20°C. Weights 5g and below are 2.7g/cm3
22 Weights
SDS
Documents