missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Economical pH Test Strips

Line of pH test strips incorporates the most up-to-date technology.

$13.82 - $13.82

Specifications

Type pH Test Strip
For Use With (Application) Stabilized to provide user with most accurate color determination
Quantity 50 Bag
View More Specs

Products 6
Catalog Number Mfr. No. pH Range Price Quantity  
Catalog Number Mfr. No. pH Range Price Quantity  
S6115
View Documents Promotion Details
Fisher Science Education
PH70101B50
7 to 10
Pack of 50 Each for $13.82
 
S6112
View Documents Promotion Details
Fisher Scientific
PH00251B50
0 to 2.5
Pack of 50 Each for $13.82
 
S6117
View Documents Promotion Details
Fisher Science Education
CZ1114
11.5 to 14
Pack of 50 Each for $13.82
 
S6111
View Documents Promotion Details
Fisher Science Education
PH00151B50
0 to 1.5
Pack of 50 Each for $13.82
 
S6113
View Documents Promotion Details
Fisher Science Education
PH30601B50
3 to 6
Pack of 50 Each for $13.82
 
S6114
View Documents Promotion Details
Fisher Science Education
PH50901B50
5 to 9
Pack of 50 Each for $13.82
 
Description

Description

 

  • Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
  • Stabilized to provide the most accurate color determinations
  • Color charts printed on each pack for quick and easy identification
  • Each pack includes 50 strips

 

Specifications

Specifications

pH Test Strip
Stabilized to provide user with most accurate color determination
50 Bag
SDS
Product Certifications