missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Economical pH Test Strips
Line of pH test strips incorporates the most up-to-date technology.
Manufacturer: Fisher Science Education CZ1114
Description
- Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
- Stabilized to provide the most accurate color determinations
- Color charts printed on each pack for quick and easy identification
- Each pack includes 50 strips
Specifications
11.5 to 14 | |
0.5 | |
pH Test Strip |
Stabilized to provide user with most accurate color determination | |
50 Bag |