missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Economical Cork Stoppers
For all of your laboratory needs.
$8.97 - $8.97
Specifications
Material | Cork |
---|---|
Product Type | Stopper |
For Use With (Application) | Ideal for sealing bottles |
Grade | Laboratory |
Catalog Number | Mfr. No. | Length (Metric) | Size | Stopper Number | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Size | Stopper Number | Price | Quantity | |||
S49193
|
Customer Channels Group (Ccg)
1XXXPK100 |
1.58 cm | 1 | 1 |
Pack of 100 Each for $8.97
|
|
|||
Description
- 15 different sizes
- Laboratory grade
Specifications
Cork | |
Stopper | |
Ideal for sealing bottles | |
Laboratory |