Please login to your online account to display your discounted pricing
Economical Allihn Condensers
$244.86 - $272.43
Specifications
Includes Spherical bulbs and sealed-in water jacket
Material Borosilicate Glass
View More Specs
Products
Catalog Number Mfr. No. Length (English) Jacket Length (Metric) Jacket Price Quantity    
07-730A
amk glass inc
TG2310001
7.8 in. 20cm Each for $244.86
07-730B
amk glass inc
TG2310003
11.8 in. 30cm Each for $272.43
Description & Specifications
Specifications
Includes Spherical bulbs and sealed-in water jacket
Material Borosilicate Glass
Water jackets sealed to condensing tubes. Bulbs approximately spherical. Use No. 3 rubber stopper. Borosilicate glass.