missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ EconoLINE Electrodes

Easy to use and low maintenance, but with reliable and dependable materials, with an excellent price-to-performance ratio. EconoLINE electrodes have a low-resistance, A type glass membrane and Ag/AgCl reference system for dependable results.

$146.69 - $146.69

Specifications

Filling Solution Type Gel
For Use With (Equipment) PY-E50-2S ; PY-B1L-2S;PY-A01-2S
Description Sensing tip shape: Cylindrical with tabs
Junction Fiber
Reference Type Ag/AgCl
View More Specs

Includes: Electrode, Manual, 50mL refill bottle

Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
14-557-228
View Documents
Sartorius™
PYP25
pH Combination Electrode (no ATC)
Each for $146.69
 
Description

Description

  • Easy to Use: no refilling required with gel-filled electrolyte
  • Easy to Clean: exterior wipedown epoxy body style
  • Protection tab reduces breakage of glass membrane from magnetic stir bars or when sampling
  • Bulb Style

Warranty

1 year

Specifications

Specifications

Gel
Sensing tip shape: Cylindrical with tabs
Ag/AgCl
NTC 10kilohm
Polysulfone Plastic
PY-E50-2S ; PY-B1L-2S;PY-A01-2S
Fiber
Ambient temperature
1 Year
SDS