missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Econazole Nitrate, TCI America™

Manufacturer:  TCI America E095725G

 View more versions of this product

Catalog No. E095725G



Specifications

Specifications

Econazole Nitrate
161°C
24169-02-6
MFCD00058160
DDXORDQKGIZAME-UHFFFAOYSA-N
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
68589
Crystal-Powder at 20°C
White
25g
C18H16Cl3N3O4
econazole nitrate, ifenec, epi-pevaryl, gyno-pevaryl, econazole nitrate salt, pevaryl, +--econazole nitrate, spectazole, econazolum nitrate, gyno-pevaryl 150
C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
444.693
444.69
≥98.0% (HPLC,T)

Chemical Identifiers

24169-02-6
444.693
DDXORDQKGIZAME-UHFFFAOYSA-N
68589
C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
C18H16Cl3N3O4
MFCD00058160
econazole nitrate, ifenec, epi-pevaryl, gyno-pevaryl, econazole nitrate salt, pevaryl, +--econazole nitrate, spectazole, econazolum nitrate, gyno-pevaryl 150
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
Safety and Handling

Safety and Handling

RTECSNumber : NI4450000

TSCA : No

Documents