Learn More
Econazole, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 27220-47-9 |
---|---|
Molecular Formula | C18H15Cl3N2O |
Molecular Weight (g/mol) | 381.68 |
MDL Number | MFCD00800993,MFCD00058160 (.HNO3) |
InChI Key | LEZWWPYKPKIXLL-UHFFFAOYNA-N |
Synonym | econazole, ecostatin, econazolum, pevaryl, gyno-pevaryl, econazol, palavale, spectazole, +--econazole, ecostatin cream |
PubChem CID | 3198 |
ChEBI | CHEBI:82873 |
IUPAC Name | 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole |
SMILES | ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AC458630050
|
Thermo Scientific Chemicals
458630050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC458630250
|
Thermo Scientific Chemicals
458630250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
27220-47-9 | |
381.68 | |
LEZWWPYKPKIXLL-UHFFFAOYNA-N | |
3198 | |
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole |
C18H15Cl3N2O | |
MFCD00800993,MFCD00058160 (.HNO3) | |
econazole, ecostatin, econazolum, pevaryl, gyno-pevaryl, econazol, palavale, spectazole, +--econazole, ecostatin cream | |
CHEBI:82873 | |
ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1 |
Specifications
27220-47-9 | |
Authentic | |
C18H15Cl3N2O | |
5g | |
Solubility in water: insoluble in water. Other solubilities: soluble in hot methanol, slightly soluble in ethanol, insoluble in ether | |
ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1 | |
381.68 | |
CHEBI:82873 | |
99% |
0.5% max. (1 g, 60°C, 4 h) | |
98.5% min. (HPLC) | |
MFCD00800993,MFCD00058160 (.HNO3) | |
econazole, ecostatin, econazolum, pevaryl, gyno-pevaryl, econazol, palavale, spectazole, +--econazole, ecostatin cream | |
LEZWWPYKPKIXLL-UHFFFAOYNA-N | |
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole | |
3198 | |
381.68 | |
Econazole |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 248-341-6
RUO â Research Use Only