missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EcoLine™ Compound Microscopes

Designed for university and laboratory use.

Manufacturer:  Fisher Scientific DELB

 View more versions of this product

Catalog No. S04755


 

Specifications Description & Specifications

Specifications

3 Years
Coaxial coarse and fine
40x, 100x, 400x, 1000x DIN
Binocular
Molded Styrofoam container with instruction brochure and Vinyl dustcover
1.25 N.A. Abbe
University
Iris
3.4V, 0.06w LED
40x,100x, 400x, 1000x DIN
110/220V
Mechanical
10X, 18mm widefield
University and laboratory use