Please login to your online account to display your discounted pricing

ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, GE Healthcare

$131.85 - $131.85

Specifications

Antigen IgG
Format Purified
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

45-001-473

 
ge healthcare biosciences
28901106
Cy3 150μg Each for $131.85
Description & Specifications

Specifications

Antigen IgG
Format Purified
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Secondary

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Western Blotting