Please login to your online account to display your discounted pricing

ECL Plex Goat anti-Mouse IgG-Cy™ 5 Secondary Antibody, GE Healthcare

$120.45 - $120.45

Specifications

Antigen IgG
Format Purified
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Monoclonal
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

45-001-210

 
ge healthcare biosciences
PA45009
Cy5 150μg Each for $120.45
Description & Specifications

Specifications

Antigen IgG
Format Purified
Host Species Goat
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Secondary

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms

Western Blotting