Please login to your online account to display your discounted pricing

Torrey Pines Scientific™ EchoTherm™ IN30 and IN40 Series Benchtop Chilling/Heating Incubators DFS Item

Chill or heat from 4° to 70°C in 27.5L or 55L volume units with precise control, even at ambient temperatures

$6,799.58 - $6,799.58

Specifications

Certifications/Compliance UL, CSA, CE
Diameter (English) Inside 10 in.
Diameter (English) Outside 18.5 in.
Diameter (Metric) Inside 64.5cm
Diameter (Metric) Outside 47cm
View More Specs
Products
Catalog Number Mfr. No. Capacity Weight (English) Height (English) Exterior Height (English) Interior Price Quantity    

11-680-35

DFS Item
 
torrey pines scientific™
IN35
0.97 cu. ft., 27.5L 36 lb. 15.5 in. 12 in. Each for $6,799.58
Description & Specifications

Specifications

Certifications/Compliance UL, CSA, CE
Diameter (English) Inside 10 in.
Diameter (English) Outside 18.5 in.
Diameter (Metric) Inside 64.5cm
Diameter (Metric) Outside 47cm
Hertz 50/60Hz
Temperature Range (Metric) 4 to 70°C
Voltage 12V
Width (English) Interior 14 in.
Width (Metric) Interior 35.6cm
Display LCD
Electrical Requirements 12V 50/60Hz
Material (Internal) ABS Plastic
Temperature Accuracy ±0.1°C
Warranty 1 Year
Width (English) 22.25 in.
Width (Metric) 56.5cm

The IN30 and IN40 Series bench-top, solid-state convection chilling/heating incubators are reliable, accurate and easy to use. Peltier-based for both heating and chilling, the units have no compressors or CFCs, making them truly energy efficient. The IN30 Series has 27.5-liter volume and the IN40 Series has 55L volume. Neither the IN30 and IN40 are programmable, however, the IN35 and IN45 are fully programmable, but can also be used in a non-programmable mode.

  • Peltier-based, no compressors, no CFCs, energy-efficient
  • Temperature range from 4° to 70°C
  • Fully programmable or non-programmable controls
  • 27.5 or 55L volumes
  • Timer with alarm and Auto-Off
  • Electronic calibration and power failure protection
  • RS-232 interface
Specifications
  • Range: 4.0° to 70.0°C (IN40/IN45 max cold is 16°C below room temperature)
  • Readability: ±0.1°C
  • Accuracy: ±0.2°C
  • Stability: ±0.1°C
  • Sensor Type: Solid State IC
  • Controller Type: PID
  • Chamber Uniformity: ±0.5°C
  • Convection Fan: Yes

  • Protein crystal growth
  • BODs
  • Culture growth above, below or at ambient
  • Enzyme reactions and deactivations
  • Incubating marine cultures below ambient
  • Ligations at 14° to 16°C
  • Hybridizations
  • Storing oocytes at 17°C
  • Storing DNA libraries

UL, CSA , and CE listed

One year