Please login to your online account to display your discounted pricing

EC 23, Tocris™

Synthetic retinoid; induces differentiation of stem cells

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material EC 23
Assay Percent Range >99%
Molecular Formula C23H24O2
Formula Weight 332.44
Synonym 4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)ethynyl)-benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

401110

 
r & d systems
RDS401110
10mg Each for $205.70

401150

 
r & d systems
RDS401150
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 332.44
Quantity 10mg
Chemical Name or Material EC 23
Assay Percent Range >99%
Molecular Formula C23H24O2
CAS 104561-41-3
Synonym 4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)ethynyl)-benzoic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C