missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ebselen

$103.64 - $103.64

Chemical Identifiers

CAS 60940-34-3
Molecular Formula C13H9NOSe
Molecular Weight (g/mol) 274.192
MDL Number MFCD00210937
InChI Key DYEFUKCXAQOFHX-UHFFFAOYSA-N
Synonym ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
PubChem CID 3194
ChEBI CHEBI:77543
IUPAC Name 2-phenyl-1,2-benzoselenazol-3-one
SMILES C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63190MA View Documents Alfa Aesar™
J63190MA
10mg Each for $103.64
 
Description

Description

Applications
Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

Solubility
Soluble in ethanol, methanol, acetone, acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

60940-34-3
274.192
DYEFUKCXAQOFHX-UHFFFAOYSA-N
3194
2-phenyl-1,2-benzoselenazol-3-one
C13H9NOSe
MFCD00210937
ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
CHEBI:77543
C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2

Specifications

White
10mg
C13H9NOSe
UN3283
ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2
274.192
CHEBI:77543
Powder
180°C to 181°C
60940-34-3
MFCD00210937
14,3486
DYEFUKCXAQOFHX-UHFFFAOYSA-N
2-phenyl-1,2-benzoselenazol-3-one
3194
274.2
Ebselen
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P304+P340-P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

RTECSNumber : DE4140750

Recommended Storage : −30° to −10°C