Please login to your online account to display your discounted pricing

EBPC, Tocris


Aldose reductase inhibitor

Quantity 10mg

Manufacturer: r & d systems  051810

 View more versions of this product

Catalog No. 051810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 261.28
Chemical Name or Material EBPC
Assay Percent Range >99%
Molecular Formula C14H15NO4
CAS 4450-98-0
Synonym Ethyl 1-benzyl-3-hydroxy-2(5H)-oxopyrrole-4-carboxylate
Recommended Storage Store at +4°C

Specifications

Chemical Name or Material EBPC
Assay Percent Range >99%
Molecular Formula C14H15NO4
Formula Weight 261.28
Synonym Ethyl 1-benzyl-3-hydroxy-2(5H)-oxopyrrole-4-carboxylate
Accessories

Store at +4°C