Please login to your online account to display your discounted pricing

EBI3 (IL-27 subunit) Rat anti-Mouse, Clone: DNT27, eBioscience™

Rat Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen EBI3 (IL-27 subunit)
Clone DNT27
Host Species Rat
Gene Alias EBI-3, Interleukin-27 subunit
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013671

 
life technologies
LS14727380
25μg Each for $129.05

5013672

 
life technologies
LS14727382
100μg Each for $332.05
Description & Specifications

Specifications

Antigen EBI3 (IL-27 subunit)
Clone DNT27
Host Species Rat
Gene Alias EBI-3, Interleukin-27 subunit
Species Reactivity Mouse
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody DNT27 reacts with mouse Epstein-Barr Virus-induced gene 3 (EBI3). EBI3 is a 34kDa soluble protein first identified in B cells that is upregulated after Epstein-Barr virus infection. EBI3 can exist as a homodimer and also heterodimerizes with IL-27α (p28, IL-30) to form the cytokine IL-27. IL-27 is produced most abundantly by dendritic cells and macrophages with some recent reports of expression in NKT cells. Recently, a complex of EBI3 and the p35 subunit of IL-12, called IL-35, has been reported to be produced by regulatory T cells.