missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ebastine, TCI America™

Manufacturer:  TCI America E09255G

 View more versions of this product

Catalog No. E09255G



Specifications

Specifications

Ebastine
87°C
90729-43-4
MFCD00865661
MJJALKDDGIKVBE-UHFFFAOYSA-N
4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
3191
Crystal-Powder at 20°C
White
5g
C32H39NO2
ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish
CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4
469.669
469.67
≥98.0% (HPLC,T)

Chemical Identifiers

90729-43-4
469.669
MJJALKDDGIKVBE-UHFFFAOYSA-N
3191
CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4
C32H39NO2
MFCD00865661
ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish
4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
Safety and Handling

Safety and Handling

RTECSNumber : EL8140000

TSCA : No

Documents