Please login to your online account to display your discounted pricing

Eaton's reagent, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369800025

 View more versions of this product

Catalog No. AC369800025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 283.89
Physical Form Liquid
Quantity 2.5L
Color Clear
Assay 6.7 to 8.2%
Density 1.5000g/mL
Fieser 05,535; 07,291; 08,400; 11,428
CAS 75-75-2
CAS 1314-56-3
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula O10P4
MDL Number MFCD00070545
Concentration 6.7 to 8.2 %
Refractive Index 1.4325 to 1.4375 (20°C, 589nm)
Packaging Glass bottle
Synonym Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid
CAS 39394-84-8
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May be corrosive to metals. Harmful if swallowed. Harmful in contact with skin. May cause respiratory irritation.
Chemical Name or Material Eaton's reagent
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material Eaton's reagent
CAS 39394-84-8
CAS 75-75-2
CAS 1314-56-3
Assay 6.7 to 8.2%
Molecular Formula O10P4
Formula Weight 283.89
MDL Number MFCD00070545
Synonym Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid
Accessories

GHS Signal Word: Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes severe skin burns and eye damage. May be corrosive to metals. Harmful if swallowed. Harmful in contact with skin. May cause respiratory irritation.

SDS & Safety Documents