missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eaton's reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 369805000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Eaton's reagent | |
39394-84-8,75-75-2,1314-56-3 | |
O10P4 | |
6.7 to 8.2 % | |
1.5000g/mL | |
283.89 | |
Glass bottle | |
1.5 | |
>110°C |
Undesignated | |
122.0°C (1.0 mmHg) | |
MFCD00070545 | |
05,535; 07,291; 08,400; 11,428 | |
Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid | |
6.7 to 8.2% | |
1.4330 to 1.4360 | |
500 mL | |
Liquid |
RUO – Research Use Only