Thermo Scientific™ Easystrip™ Plus Tube Strip with Attached Ultra Clear Caps
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays.
$788.85 - $788.85
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB2005 Thermo Scientific™
AB2005
250 strips Case for $788.85
Specifications
Description
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays. The tube strip is compatible with 0.2mL thermal cycler blocks and features individually attached optical caps for quick and easy access to individual samples. In addition, graduated 20µL markers on each tube enable visual volume verification and measurement of reagent and sample additions.
For tube strips with flat caps, please see Cat. No. AB-2000.
Features:
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20μL markings on each tube
  • Track samples easily using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
Applications
  • qPCR/Real-Time PCR
For Research Use Only. Not for use in diagnostic procedures.