Please login to your online account to display your discounted pricing
Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination in PCR assays.
$695.33 - $695.33
Products
Catalog Number Mfr. No. Quantity Price Quantity    
AB2000
thermo scientific™
AB2000
250 Strips Case for $695.33
Description & Specifications
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination in PCR assays. The tube strip is compatible with 0.2mL thermal cycler blocks and features individually attached flat caps for quick and easy access to individual samples. In addition, graduated 20µL markers on each tube enable visual volume verification and measurement of reagent and sample additions.
For tube strips with ultra clear caps, see Cat. No. AB-2005.
Features:
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20μL markings on each tube
  • Track samples using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
Applications:
  • PCR