Please login to your online account to display your discounted pricing
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
$241.23 - $241.23
Specifications
Volume (Metric) 0.2mL
Reaction Volume 15 to 20µL
Storage Requirements Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    
21-402-51
thermo scientific™
MB6228
EasyStart Micro 20 Strips Pack of 96 Each for $241.23
Case of 10 Pack for $2,412.30
Description & Specifications
Specifications
Volume (Metric) 0.2mL
Reaction Volume 15 to 20µL
Storage Requirements Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Features:
  • Delivers better specificity and yield than conventionally prepared mixtures
  • Compatible with a wide range of enzymes
  • Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
Ordering Information:
Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.