Thermo Scientific™ EASY-Spray™ LC Columns
Nano and capillary flow LC-MS proteomics applications. Unique plug-and-spray columns provide optimized, state-of-the art performance and ease of use.
Manufacturer:  Thermo Scientific™ ES810
 View more versions of this product
Catalog No. ES810
Description
Description
Ensure robust nano and capillary flow LC-MS with Thermo Scientific™ EASY-Spray™ LC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigourously tested to ensure maximum quality, these columns are designed for simplicity and ease-of-use.
Specifications
Specifications
Analytical Column, EASY-Spray
200 µm
300bar
Monolith
80°C
Reversed Phase
250 mm
Monolith Polymer
1 to 10
Phenyl
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.