Please login to your online account to display your discounted pricing

Easy Science Demos & Labs Series

Great, inexpensive demonstrations for science laboratories.

Manufacturer: fisher science education  0-45023

 View all options for this product

Catalog No. S67867


Description & Specifications


Author Thomas Kardos
Category Physics
Format Book
Grade 8 to 12
Product Type Physics Book

  • Demonstrations and labs that stimulate scientific inquiry
  • All activities have been checked for safety
  • Easy and inexpensive to perform
  • Topics covered: cells, genetics, human biology, plant and animal life
  • Grades 8 to 12
  • Thomas Kardos; Paperback; Second Edition, 2004; 116 pages