Please login to your online account to display your discounted pricing

Easy Science Demos & Labs Series


Great, inexpensive demonstrations for science laboratories.

Manufacturer: fisher science education  0-4499X

 View more versions of this product

Catalog No. S67865

  • / Each
 

Description & Specifications

Specifications

Format Book
Grade 8 to 12
Product Type Chemistry Book
Author Thomas Kardos
Category Chemistry