Please login to your online account to display your discounted pricing

Easy Science Demos & Labs Series


Great, inexpensive demonstrations for science laboratories.

Manufacturer: fisher science education  0-45015

 View more versions of this product

Catalog No. S67864

  • / Each
 

Description & Specifications

Specifications

Product Type Life Science Book
Author Thomas Kardos
Category Science
Format Book
Grade 8 to 12