Please login to your online account to display your discounted pricing

Easy Science Demos & Labs Series

Great, inexpensive demonstrations for science laboratories.

$32.40 - $32.40

Specifications

Author Thomas Kardos
Format Book
Grade 8 to 12
View More Specs
Products
Catalog Number Mfr. No. Category Product Type Price Quantity    

S67865

 
fisher science education
0-4499X
Chemistry Chemistry Book Each for $32.40

S67864

 
fisher science education
0-45015
Science Life Science Book Each for $32.40

S67866

 
fisher science education
0-45007
Geography Earth Science Book Each for $32.40
Description & Specifications

Specifications

Author Thomas Kardos
Format Book
Grade 8 to 12

  • Demonstrations and labs that stimulate scientific inquiry
  • All activities have been checked for safety
  • Easy and inexpensive to perform
  • Topics covered: cells, genetics, human biology, plant and animal life
  • Grades 8 to 12
  • Thomas Kardos; Paperback; Second Edition, 2004; 116 pages