Please login to your online account to display your discounted pricing

Easy Science Demos & Labs Series


Great, inexpensive demonstrations for science laboratories.

Manufacturer: fisher science education  045031-906

 View more versions of this product

Catalog No. S68765

  • / Each
 

Description & Specifications

Specifications

Grade 8 to 12
Author Thomas Kardos
Format Book
Product Type Easy Science Demos and Labs: Complete Series
For Use With (Application) Great, inexpensive demonstrations for science laboratories
Category Science