missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B™ Instrument Easy-Read™ General Purpose Liquid-In-Glass Thermometers

Easy-to-read partial immersion thermometers.

Manufacturer:  Fisher Science Education B603020100

 View more versions of this product

Catalog No. S95794



 

Specifications

Description

Thermometer, General Purpose; H-B Instrument; Easy-Read; Liquid-In-Glass; Enviro-Safe liquid; Range: 20deg. to 500deg.F; Accry: +/-2F, +/-3F above 230F, +/-4F above 392F; Partial Immersion: 76mm; Div.: 2deg.F; Length: 350mm

  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Description & Specifications

Specifications

±2°F, ±3°F above 230°F, ±4°F above 392°F
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial
14 in.
Yellow Back Glass
20° to 500°F
Enviro-Safe™
Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
76mm (3 in.)
Multilanguage instructions, statement of accuracy
350mm
2°F
Not Applicable
Promotions

Promotions